Ritmed DisTech™ Earloop Face Masks, Medicom
Catalog #: CA76474-114
Supplier: Medicom
Specifications
- Sterility:Non sterile
- Color:Blue
- Description:DisTech™ earloop face mask with earloops, ASTM Level 2
- Cat. no.:CA76474-114
Specifications
About this item
Reliable ASTM Level 2 protection and exceptional value from the mask experts.
- Shingle pleat design prevents fluid pooling and cross-contamination
- Adjustable nosepiece provides a personalized fit
- Quality materials ensure enhanced breathability
- Ultrasonically sealed, soft, stretchy, latex-free earloops ensure wearer comfort
These ASTM Level 2 masks are ideal for lower-risk procedures, providing reliable protection while preventing the risk of cross-contamination.
Packaging: Masks are packaged 50/box, 10 boxes/case.
Customers Who Bought This Also Bought