AssureTouch™ Ultra Tough Nitrile Gloves
Supplier: Avantor
New Product
Certificates
About this item
Extra protection and grip when needed.
- Textured fingertips ensure a secure grip, even when wet
- Ambidextrous design
- Powder-free and latex-free
Industrial quality nitrile ensures high chemical and tear resistance without compromising comfort or fit.
Packaging: Gloves are packaged 100 gloves/box, 10 boxes/case (1000 gloves/case).
Specifications
- Catalog No:
- CA76552-258
- CA76552-244
- CA76552-256
- CA76552-250
- CA76550-127
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Fully textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded