AssureTouch™ Pure Nitrile Gloves
Supplier: Medicom
New Product
Certificates
About this item
Strong protection that’s safe for skin.
- Ultra-thin nitrile gloves are ideal for procedures that require precision and tactile sensitivity
- Textured fingertips ensure a secure grip, even when wet
- Chemo-tested
This thin nitrile glove is ideal for procedures that require a high level of precision and tactile sensitivity. DermaSense technology helps reduce the risk of skin reactions that could be caused by chemical accelerators commonly found in nitrile gloves.
Packaging: Gloves are packaged 200 gloves/box, 10 boxes/case (2000 gloves/case).
Specifications
- Catalog No:
- CA76579-725
- CA76579-723
- CA76579-717
- CA76579-721
- CA76579-719
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded