AssureTouch® Agile™ Nitrile Medical Examination Gloves
Supplier: Medicom
Certificates
About this item
Mobility and protection when it counts.
- Textured fingertips
- Chemo tested
- Comfortable fit
- Optimal stretch
- Latex-free
These nitrile gloves offer superior puncture, tear, and chemical resistance without compromising dexterity or tactility. Textured fingertips ensure a secure grip, even when wet.
Packaging: Gloves are packaged 100 gloves/box; 10 boxes/case (1000 gloves/case).
Specifications
- Catalog No:
- CA76550-132
- CA76550-130
- CA76550-142
- CA76550-140
- CA76550-124
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Finger textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded