Specifications
- Description:Multi-sample osmometer
- Cat. no.:76579-778
Specifications
About this item
Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality, and purity.
- Multi-sample osmometer
- Gold standard freezing point technology
- Fully compliant with 21 CFR part 11, GMP, and EU Annex 11 with e-Signature
- Meets Pharmacopeia osmolality testing guideline
Designed to meet the high sample throughput, regulatory compliance (including 21 CFR part 11), and data management needs of busy bioprocessing and biomanufacturing labs, the OsmoTECH PRO Multi-Sample Micro-Osmometer combines ease-of-use with accurate and precise osmolality measurements for reliable and reproducible results. Optimize and ensure the quality and consistency of buffers, media, product, excipients, and process parameters to maximize biologic yield, quality and purity.