Specifications
- Description:Single-sample osmometer
- Cat. no.:76579-780
Specifications
About this item
Increase your productivity and reliability with powerful freezing point depression technology for osmolality measurement with the OsmoTECH® XT single-sample osmometer.
- Offers the widest range of osmolality testing (0 to 4000 mOsm/kg H₂O)
- Large user-friendly interface
- New, intelligent freezing technology
- Fully compliant with 21 CFR part 11, GMP, and EU Annex 11 with e-Signature
- Meets Pharmacopeia osmolality testing guidelines
The OsmoTECH® XT measures the widest variety of sample types (high concentration, high viscosity) accurately across bioprocessing and cell and gene therapy workflows. It’s also designed to make your work as streamlined and comfortable as possible with even more robust data integrity and compliance features to match your digital environment. See how the OsmoTECH® XT osmometer will change the way you work in the lab.
OsmoTECH® XT Single-Sample Micro-Osmometer Installation and Training (sold separately): Our installation and training experts ensure that your instrument is properly installed and your staff is properly trained. Training topics include installation, operation, and maintenance. Benefits of installation and training include: proper installation and setup, unpacking and inspection, Instrument training for staff, Inspector-ready training documentation. Our installation and training experts ensure that your instrument is properly installed and your staff is properly trained.