Kugelmeiers Spherical plate 5D Three-Dimensional Cell Culture Plate, Heidolph
Catalog #: 76583-036
Supplier: Heidolph NA, LLC
Specifications
- Sterility:Sterile
- Color:Clear and transparent
- Description:Heidolph 3D cell culture plates, 9000 spheroids per plate, 750 microwells per well
- Material:Cyclic olefin copolymer (COC)
Specifications
About this item
The spherical plate 5D is a sterilized 3D cell culture plate that can produce up to 9000 uniform, size-controlled, standardized spheroids.
- Standard 24 well plate dimension (12 wells have microwells (3D spheroid production), 12 wells do not have microwells (control))
- Each well has 750 microwells, allowing the plate to produce up to 9000 spheroids
- Plates are sterilized and have pre-applied anti-adherence coating, making them ready to use out of the box
- There are only three main steps in the SOP and it just takes 24 hours for your 9000 spheroids to be produced
- A wide range of cell lines, islet cells, and stem cells have been used on this plate successfully
The patented geometry of the microwell standardizes spheroid production: the spheroids are uniform with other spheroids and the individual spheroids have almost perfect spherical dimensions.
Certifications: Guaranteed sterility assurance level (SAL) 10⁻⁶ according to ISO 11137.
Customers Who Bought This Also Bought