Eisco Glass Wide Neck Erlenmeyer Flasks
Supplier: Wards
Certificates
About this item
Wide Neck borosilicate glass Erlenmeyer flasks with beaded rims and white graduations.
- Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
- Wide neck for easier filling and cleaning
- Chip and crack resistant, fire polished beaded rims
- Easy-to-read white enamel graduations and specifications for contrast against dark liquids
Wide neck Erlenmeyer flasks are made of high quality borosilicate 3.3 glass. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
- Strain point 515 °C
- Annealing point 565 °C
- Softening point 820 °C
Customers Who Bought This Also Bought