FlexTech™ Y9298 Cut-Resistant Polyurethane Palm Coated Gloves, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Protect hands with ANSI A4 cut resistance and a polyurethane palm.
- 18 gauge thin glove for better dexterity
- Touchscreen compatible
Palm coated work gloves provide additional protection in high wear/high risk areas of the wearer's hand. The Y9298 glove series features an ultra-lightweight liner that provides excellent dexterity with ANSI Cut level A4 protection to reduce hand injuries, while providing increased abrasion resistance and durability for prolonged use.
Certifications: ANSI A4 cut resistance. ANSI puncture level 4. ANSI Abrasion level 4.
Specifications
- Catalog No:
- 76595-894
- 76595-892
- 76595-898
- 76595-904
- 76595-896
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 18
- 18
- 18
- 18
- 18
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile