Funnel, Fluted, 60° Angle, Long Stem, PYREX®, Corning
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°. These funnels are fabricated from blanks formed to a 60° angle, which permits accurate fitting of filter paper and thus reducing filtering time. In addition, few funnels have depressed inside flutings which further decrease filtering time.
Specifications
- Catalog No:
- 30232-061
- 30232-050
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey