PYREX® Funnel, Plain, 60° Angle, Short Stem, Large Size
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.
Specifications
- Catalog No:
- 89090-548
- 89090-544
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey