PYREX® Funnels with Long Stem, Corning
Supplier: Corning
Certificates
About this item
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With a beaded edge for strength. Cone angle 60°.
These funnels are for use in flasks or test tubes without a filter rack. Fluted funnels feature pressed inside flutings to further decrease filtering time.
Specifications
- Catalog No:
- 89090-546
- 89091-640
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey