Corning® Clear Bottom Black and White Polystyrene Microplates, 96 Well, Corning
Supplier: Corning
Certificates
About this item
PS, flat bottom.
- Bottoms are 60% thinner than conventional PS plates, resulting in lower background fluorescence and enabling readings down to 340 nm
- Opaque walls prevent well-to-well crosstalk
- Optically clear flat bottom profile permits direct microscope viewing
NBS™ = Non Binding Surface.
Specifications
- Catalog No:
- 33501-008
- 89184-616
- 33501-004
- 33501-006
- 33501-820
- 89091-010
- 33501-818
- 89092-710
- 33500-998
- 33501-002
- 29444-010
- 89091-012
- 89091-014
- 29444-008
- 29444-037
- 33500-994
- 33500-996
- 29444-332
- 29444-330
- 29444-099
- 66025-630
- 89091-018
- 89092-708
- 29445-162
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
Specifications
Related Information
Certificates
Search by Lot Number29444-008_14319010 29444-008_32419018 29444-008_32719004 29444-330_33919054 29444-332_08619011
Conformity declaration
More
Corning New Lab Startup Program
Promotion Material
Customers Who Bought This Also Bought