Corning® BioCoat® Collagen I-Coated Flasks, Corning
Supplier: Corning
Certificates
About this item
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. In vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
- Promotion of cell attachment and spreading
- Rapid expansion of cell populations
- Serum-free or reduced serum culture
- Cell adhesion assays
- Improving survival of primary cells in culture
- Storage: 4 to 30 °C under dry conditions
Related Information
Certificates
Search by Lot Number12777-074_27019013 12777-080_08119008 12777-080_31219013 12777-082_27019016 62405-025_13019011
Conformity declaration
More
Customers Who Bought This Also Bought