Corning® 96-Well Polyvinyl Chloride (PVC) Microplates
Catalog #: 29442-384
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Well Volume:250 µL
- Description:Nonsterile Plate without Lid
- Material:Flexible Vinyl
- Cat. no.:29442-384
- Packaging:25/Bag
- Well Shape:Round-Bottom
- Surface Treatment:Untreated
- Supplier No.:2797
Specifications
About this item
Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
- Well volume of 250 µl (260 µl for V-bottom)
- Working well volume of 50 µl to 150 µl
Ordering information: Lids are not available.
Customers Who Bought This Also Bought