Order Entry
ContactUsLinkComponent
NanoPhotometer® C40-TOUCH UV/Visible Spectrophotometer for Standard Cuvette Applications, Implen
NanoPhotometer® C40-TOUCH UV/Visible Spectrophotometer for Standard Cuvette Applications, Implen
Catalog #: CA10770-496
Supplier:  IMPLEN, INC. CA
cUL Listed
CSA Certified
UL Listed
Spectrophotometer
NanoPhotometer® C40-TOUCH UV/Visible Spectrophotometer for Standard Cuvette Applications, Implen
Catalog #: CA10770-496
Supplier:  IMPLEN, INC. CA
cUL Listed
CSA Certified
UL Listed

Specifications

  • Detector:
    4096 CMOS Array
  • Light source:
    Xenon flash lamp
  • Memory:
    32 GB
  • Sample size:
    Depends on cuvette
  • Description:
    NanoPhotometer® C40-TOUCH UV/visible spectrophotometer for standard cuvette applications
  • Wavelength reproducibility:
    ±0.2 nm
  • Wavelength accuracy:
    ±0.75 nm
  • Detection limit:
    Detection range BSA: 0.003 - 3.7 mg/ml (cuvette)
    Detection range dsDNA: 0.1 - 130 ng/µl (cuvette)
  • Absorbance accuracy:
    <1.75 % @ 0.7 A @ 280 nm
  • Compatible with:
    Windows 8, 10 (32 and 64 bit), OS X (Intel x86 and Apple M1), iOS and Android OS
  • Dimensions:
    20×20 cm
  • Display:
    1024×600 pixels; glove compatible touchscreen
  • Interfaces:
    2x USB A, USB B, HDMI, Ethernet, WiFi
  • Languages:
    English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
  • Photometric range:
    0 - 2.6 A
  • Reproducibility:
    <0.002 A @ 0 - 0.3 A @ 280 nm
    CV <1% @ 0.3 - 2.0 A @ 280 nm (Absorbance)
  • Response time:
    2.5 s measurement time
  • Sample compartment:
    1 sample capacity
  • Spectral bandwidth:
    <1.8 nm
  • Weight:
    <5.2 kg
  • Spectral Range:
    200 - 900 nm spectrum scan
  • For Use With:
    Nucleic acids, protein UV, protein assays, kinetics, OD600, wavelength, wavescan, standard curve, absorbance ratio, concentration, custom
  • Supplier No.:
    C40-TOUCH

Specifications

About this item

Implen has become the leading expert for innovative, high-quality spectroscopy instruments and the NanoPhotometer® is trusted by thousands of researchers worldwide.

  • Preprogrammed applications for (labeled) DNA, RNA and oligos (260 nm)
  • (Labeled) purified proteins and peptides (200 to 280 nm)
  • Protein assays (BCA 562 nm, Bradford 595 nm, Lowry 650 nn)
  • Kinetics (200 to 900 nm)
  • Optical density measurements (600 nm)
  • Manual methods (wave length, wave scan, ratio, concentration, standard curve)
  • Recalibration-free
  • Built-in cuvette
  • Optional CFR21 software available
  • Optional IQOQ package available

The flexible Implen NanoPhotometer® C40 UV/VIS spectrophotometer offers cuvette measurements, made in Germany. It provides full scan data from 200 - 900 nm for various life science applications like quantification and qualification of nucleic acids, proteins or cell cultures. The stand-alone design with an integrated touchscreen and small footprint make it ideal for bench operation in pharma, biotech or academic laboratories. Quantify and qualify DNA, RNA, protein, small molecules, nano particles and other samples using all kinds of standard cuvettes (plastic, quartz and other materials) and have full-spectral data in only 2.5 s.

User-friendly and secure Linux-based software with intuitive interface ensure that even unexperienced users can work with the instrument without extensive training. Stand-alone design eliminates the need for a separate computer saving bench space and and bypasses possible problems with future windows updates, however, external computers, tablets and smartphones can be connected if desired. Data export as secure id's file or full excel/PDF including graph and absorbance data for further data analysis, printing and archiving.

Recalibration-free design due to True Path Technology™ saves money for costly maintenance and ensures peace of mind over the entire life time. The NanoPhotometer® tests itself during every start of the instrument, providing feedback to the user that everything works fine. No reconditioning of the pedestal required guarantees accurate and stable performance.

Integrated Sample Control™ and Blank Control™ for analysis of sample quality and identification of contaminants via ratios and spectral analysis make sure you work with good samples only, improving success of downstream experiments like (q)PCR, cloning, micro-array, drug production and protein purification. Impure samples are flagged for immediate identification and possible contaminants are listed.

Robust design of metal and glass parts mean you can disinfect the entire device using 70% ethanol. Built-in file server and LIMS/eLN integration through REST API offering future-proof data export via USB and ethernet cable or WiFi. Barcode reader and dymolabel printer ready.

Certifications: ISO 9001:2015, CE, IEC 61010-1:2012 and EN 61326-1:2013.
Ordering information: Optional control via Windows PC, Mac, tablet and smart phone (Android and iOS). Optional built-in battery (not included) enables mobile operation without cables under fume hoods, lamina flows or sharing the device between labs. Optional CFR21 software available for US FDA 21 CFR part 11 compliance in GxP regulated environment. Optional IQOQ is available including documents, standard filters and solutions to validate your device. Contact your Avantor representative for more information.
Delivery: Including power supply, USB cable, dust cover, software and manual.
Customers Who Bought This Also Bought