Specifications
- Compatible with:R10.4.1 flow cells and kit 14 chemistry
- Description:Native barcoding auxiliary kit v14
- Cat. no.:76595-412
- No. of Reactions:12
Specifications
About this item
This expansion is recommended for users who want to make use of unused barcodes to maximise the use of their Native Barcoding Kits.
This expansion pack contains additional library preparation reagents for the Native Barcoding Kit 24 and 96 V14 (SQK-NBD114.24 and SQK-NBD114.96) for another 12 reactions in conjunction with the Sequencing Auxiliary Vials V14 (EXP-AUX003).
The Native Barcoding Auxiliary Kit V14 (EXP-NBA114) can only be used with Kit 14 chemistry. This kit is not compatible with previous kits chemistries.