Specifications
- Sterility:Sterile
- W×L:200×200 mm
- Description:Ready-to-use wipes
- Packaging:Pouch, 20×15 wipes,
Specifications
About this item
Klerwipe™ Neutral Detergent Pouch Wipes are sterile non-woven, low particulate wipes impregnated with a neutral, non-ionic detergent in a convenient resealable pouch.
- Made from hydroentangled polyester/cellulose
- High-Performance Residue Removal: Klercide™ Neutral Detergent effectively removes soils and residues as part of daily cleaning, as well as following excursions and shutdowns
- Ready-to-Use Pouch Wipes: Pre-saturated neutral detergent wipes are ready to use and ideal for streamlining compliant cleaning of equipment, bench tops and other smaller surfaces in the cleanroom
- Premium-Grade Substrate: High-quality polyester-cellulose wipes are lowlinting and prelaundered in an ISO Class 4 cleanroom to significantly reduce particles
- Sterile Production: Wipes are pre-saturated with 0.2 micron-filtered detergent, ultrasonically sealed and gamma irradiated in a Grade C/ISO Class 7 cleanroom; supported with certificates of analysis and Certificates of Sterility with every batch
- Convenient Packaging: Ready-to-use wipes come in 15-wipe packs to reduce waste and come double-bagged to simplify movement into high-grade areas, with Kleartear for easy opening with gloved hands
For cleaning small, hard cleanroom surfaces, Klerwipe™ Neutral Detergent Pouch Wipes are ideal for routine use. Effective on oily or sticky spillages or following general maintenance, the hydroentangled polyester/cellulose wipes are presaturated with Klercide™ Neutral Detergent and Water for Injection (WFI). Effectively and efficiently remove soils and residues from small surfaces in the cleanroom with ready-to-use pouch wipes pre-saturated with Klercide™ Neutral Detergent.
Ready-to-use, quick and easy to apply, Klerwipe Neutral Detergent wipes are impregnated with Klercide Neutral Detergent, a low-foaming, non-ionic solution with neutral pH. These non-corrosive wipes are powerful and effective in a wide range of cleanroom applications.
Basis weight: 58-78 g/m²