Quantum Levitation Starter Kit
Catalog #: 470346-496
Supplier: QUANTUM LEVITATION LTD
Specifications
- Description:Maglev starter kit
- Cat. no.:470346-496
Specifications
About this item
The starter kit is the fundamental, entry level kit for classroom demonstration of quantum levitation and flux pinning. Perfect for high-school students as well as university undergraduates.
- The kit is easy to use
- Highly durable and portable
A unique experimentation platform to demonstrate frictionless motion and quantum phenomena such as the Meissner effect and flux pinning.
Delivery: The starter kit comes with handheld magnetic device, medium/large quantum levitator (various flavors available), plastic tweezers, and user manual.
Customers Who Bought This Also Bought