Quantum Levitation Force Experiment
Quantum level forces experiment
Catalog # 470346-494
Supplier: QUANTUM EXPERIENCE LTD
Supplier #: 410-112
Jump to:
Product Details & Documents
Explore the magnetic forces between magnets, superconductors and metals.
- The Meissner effect
- Flux pinning
- Magnetic forces
- Letz' law
Visualize and measure the magnetic forces that Quantum Levitation so unique and unintuitive - see how a superconductor is repelled by a magnet (positive force - Meissner effect) and after it has been locked, it is attracted (negative forces - flux pinning) to it.
Documents
Specifications
- Description:Quantum level forces experiment