Methanol ≥99.9% ACS for HPLC
Catalog #: BJCS230AH200
Supplier: Honeywell Research Chemicals
CAS Number: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Stainless steel drum, returnable
- Size:200 l
- Cat. no.:BJCS230AH200
- Supplier No.:CS230AH200
Specifications
Specification Test Results
- Methanol ≥99.9% ACS for HPLC:
Specification Test Results
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
:Storage temperature: Ambient
Boiling Pt:64.7 °C
Melting Pt:–98 °C
Density:0.792 g/cm3
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.7 °C
:Melting Pt:
:Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Formula:CH₃OH
MW:32.04 g/mol
EINECS:200-659-6
:64.7 °C
Storage Temperature:Ambient
MDL Number:MFCD00004595
CAS Number:67-56-1
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS08
:GHS02
Customers Who Bought This Also Bought