Evergreen Oro-Col™, Caplugs Evergreen
Catalog #: 76499-448
Supplier: Caplugs
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Color:Natural
- Description:Oro-Col™
- Material:PP
Specifications
About this item
Oro-Col™ is an oral fluid collection system used primarily for drug abuse and hormone testing. Self-contained device for the direct collection and transport of oral fluid.
- Simple in design is convenient to use
- Consisting of a collection funnel and lid, calibrated vial and a screw cap
- Collection funnel reduces the chance of contamination
- All polypropylene construction
Customers Who Bought This Also Bought