VWR® Hazardous Location (Explosion Proof) Laboratory Refrigerators Standard Series with Natural Refrigerants
Supplier: VWR International
Certificates
About this item
These laboratory refrigerators are engineered for safety in potentially hazardous laboratory conditions.
- Microprocessor temperature controller
- Keyed door lock
- ENERGY STAR certified*
They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
All units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
Material: White powder coat
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines, ETL listed.
*Select model ENERGY STAR certified: 76579-138
*Select model ENERGY STAR certified: 76579-138
Ordering information: All units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.
Specifications
- Catalog No:
- 76579-136
- 76579-138
- 76579-140
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76579-136
- 76579-138
- 76579-140
- Defrost:
- Cycle
- Cycle
- Cycle
- Shelves/drawers:
- 5 adjustable shelves
- 5 adjustable shelves
- 5 adjustable shelves
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Electrical:
- 115 V, 60 Hz, 3 A, ⅕ HP
- 115 V, 60 Hz, 3 A, ¼ HP
- 115 V, 60 Hz, 3 A, ¼ HP
- Weight:
- 91 kg (200 lbs.)
- 108 kg (240 lbs.)
- 118 kg (260 lbs.)
- Environmental certification:
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
Specifications
Related Information
TDS Standard Series Hazardous Location (Explosion Proof) Refrigerator VWR-HC-ERP-14 76579-136.pdf TDS Standard Series Hazardous Location (Explosion Proof) Refrigerator VWR-HC-ERP-17 76579-138.pdf TDS Standard Series Hazardous Location (Explosion Proof) Refrigerator VWR-HC-ERP-20 76579-140.pdf TDS VWR Standard Series Hazardous Location Refrigerator, 14 cu. ft. TDS VWR Standard Series Hazardous Location Refrigerator, 17 cu. ft.
Datasheet
More
Customers Who Bought This Also Bought