VWR® Hazardous Location (Explosion Proof) Laboratory Freezers Manual Defrost Standard Series with Natural Refrigerants
Supplier: VWR International
Certificates
About this item
These laboratory freezers are engineered for safety in potentially hazardous laboratory conditions.
- Microprocessor temperature controller
- Keyed door lock
- ENERGY STAR certified*
They utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist. For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see flammable material storage units.
All units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
Material: White powder coat
Certifications: These units meet NFPA and OSHA guidelines for hazardous location, Class 1, Division II, Groups C and D guidelines, ETL listed.
*Select models ENERGY STAR certified: 76579-130, 76579-132, and 76579-134
*Select models ENERGY STAR certified: 76579-130, 76579-132, and 76579-134
Ordering information: All units come with a two year parts and labor warranty.
Caution: Units are not equipped with a line cord and plug. They must be hardwired in conduit to 120 VAC power source for safety.
Specifications
- Catalog No:
- 76579-134
- 76579-130
- 76579-132
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76579-134
- 76579-130
- 76579-132
- Defrost:
- Manual
- Manual
- Manual
- Shelves/drawers:
- 6 fixed shelves
- 6 fixed shelves
- 6 fixed shelves
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Electrical:
- 115 V, 60 Hz, 6 A, ½ HP
- 115 V, 60 Hz, 3.5 A, ⅓ HP
- 115 V, 60 Hz, 6 A, ½ HP
- Weight:
- 109 kg (240 lbs.)
- 88 kg (195 lbs.)
- 100 kg (220 lbs.)
- Environmental certification:
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
- Units use natural, hydrocarbon refrigerants
Specifications
Related Information
TDS Standard Series Hazardous Location (Explosion Proof) Freezer VWR-HC-EFP-14 76579-130.pdf TDS Standard Series Hazardous Location (Explosion Proof) Freezer VWR-HC-EFP-17 76579-132.pdf TDS Standard Series Hazardous Location (Explosion Proof) Freezer VWR-HC-EFP-20 76579-134.pdf TDS VWR Standard Series Hazardous Location Manual Defrost Freezer, 14 cu. ft. TDS VWR Standard Series Hazardous Location Manual Defrost Freezer, 17 cu. ft.
Datasheet
More
Customers Who Bought This Also Bought