ContactUsLinkComponent
Fluorescence Stereomicroscope, MZ10F, Leica Microsystems
Fluorescence Stereomicroscope, MZ10F, Leica Microsystems
Catalog #: CA76575-544
Supplier:  Leica Microsystems
Fluorescence Stereomicroscope, MZ10F, Leica Microsystems
Catalog #: CA76575-544
Supplier:  Leica Microsystems
Specifications
  • Magnification:
    8× - 80×
  • Description:
    MZ10F Fluorescence stereomicroscope
  • Cat. no.:
    CA76575-544
Specifications
About this item
Modular stereo microscope for fluorescent imaging.
  • Magnification: 8× to 80×
  • 375 Lp/mm high resolution
  • Triple beam technology
  • Fluo III rapid filter changer
The Leica MZ10F is the ideal tool for fluorescence applications such as gene expression studies or developmental biology applications. Leica's Triple Beam technology designates a separate beam path specifically for fluorescent illumination, resulting in high contrast and detailed fluorescent imaging. The Fluo III rapid filter changer allows for fast interchange between four filter sets, for easy convenient imaging.
Ordering information: For more information on the MZ10F, please contact your local Avantor representative.
Delivery: MZ10F package includes optics carrier, TL3000 Ergo transmitted light base, 1.0× PLAN APO objective lens, LED3 fluorescence light source, ET GFP filter set, LED5000 Spot Light Illuminators.
Customers Who Bought This Also Bought