EZwaste® UN/DOT Tight Head Containers for Shipping, Transport, Storage, and Waste, UN 3H1, Foxx Life Sciences
Supplier: Foxx Life Sciences
Certificates
About this item
The UN/DOT solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system eliminated satellite collection and waste room consolidation.
- Material HDPE
- 10L UN Rating: 3H1/Y1.8/100
- 60L UN Rating: 3H1/Y1.8/150
HDPE UN/DOT system is designed to be an affordable solution for liquid waste disposal. The containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 13.5, 20, and 60 L sizes. The 10 L UN/DOT containers come with the 50S cap and the 13.5 L through 60 L UN/DOT containers come with the S70 Cap.
Certifications: UN 3H1 certified.
Customers Who Bought This Also Bought