Eisco Bunsen Burner
Supplier: Wards
Certificates
About this item
Economical option for general laboratory applications.
- 11 mm burner tube does not have a flame stabilizer
- Serrated inlet arm that accepts 6 to 8 mm tubing
- Mounted on a stable, circular base
Economical, laboratory quality gas burner ideal for general lab applications. Burner comes equipped with a serrated inlet arm that measures 8 mm in Ø and 1" in length. No flame stabilizer or gas adjustment. Burner tube Ø is 13 mm.
Overall size: 5.7tall×3" Ø base.
Customers Who Bought This Also Bought