Cryo Racks, United Scientific Supplies
Supplier: United Scientific Supplies
Certificates
About this item
Strong polycarbonate boxes for storage of cryogenic vials.
- Autoclavable
- Polycarbonate
- Temperatures range: –196 to +121 °C
Manufactured of strong polycarbonate, these boxes are ideal for compact storage of 2.0 or 5.0 ml cryogenic vials. Boxes can be used at temperatures from –196 to 121 °C and are autoclavable. Transparent lids have a numbered grid for sample identification. Frosted surface area is suitable for writing with markers. Dimensions of cryo boxes are designed to be compatible with vertical, horizontal, and drawer style freezer racks. 66301 includes two red boxes, one blue box, and one yellow box. 66302 includes two red boxes and two blue boxes. 66303 includes an assortment of red, yellow, and blue colored boxes.
Customers Who Bought This Also Bought