CapSure®–LP Edge Bordered Sealed Edge Cleanroom Laundered Wipers, 100% Polyester Knit, Berkshire
Supplier: Berkshire
Certificates
About this item
CapSure®–LP Edge is a cleanroom laundered wiper.
- Patented surface treatment captures and retains particulate contamination
- 100% continuous filament polyester knit
- 4 - side sealed border
- Laundered and packaged in Berkshire’s ISO Class 4 cleanroom
- Excellent combination of cleanliness, absorbency, and abrasion resistance
- Steam autoclavable for use in aseptic environments
- Captures 42 times more particles than competitive (untreated) wipers
- Critically low particles, fibers, and extractables
CapSure®–LP Edge is a 4 - side sealed edge cleanroom laundered wiper that has been specially engineered for superior surface cleaning ability. A patented (#8,431,497) surface treatment allows the wiper to capture and retain particulate contamination, resulting in more efficient cleaning and reduced likelihood of cross contamination of critical surfaces.
CapSure®–LP Edge has the high level of cleanliness and abrasion resistance required for ISO Class 3 and above environments and applications requiring critical control of contaminants.
Certifications: ISO class 3 - 5. Cleanroom class 1 - 100.
Packaging: Stacked wipers are packaged 150 sheets/pack, 8 packs/case. Value Pack (bulk) wipers are packaged 150 sheets/pack, 8 packs/case.
Customers Who Bought This Also Bought