Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: Honeywell Research Chemicals
Certificates
About this item
Synonyms: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
Formula:H₃CCN
MW:41.05 g/mol
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:-46 °C
Density:0.7822 g/cm³ (20 °C)
MDL Number:MFCD00001878
CAS Number:75-05-8
:Storage temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:6 °C
:Boiling Pt:
:Boiling Pt: 82 °C (1.013 hPa)
:Melting Pt:
:Melting Pt: -46 °C
:Flash Pt:
:Flash Pt: 6 °C
:M.W. 41.05 g/mol
:Acetonitrile
Storage Temperature:Ambient
EINECS:200-835-2
:82 °C (1.013 hPa)
UN:1648
REACH:01-2119471307-38
:3
:II
:6 °C
:GHS07
:GHS08
:GHS02
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GC Analysis:99.9 % min.
- Water by Karl Fischer Titration:0.005 % max.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- UV Absorbance @ 200 nm:0.05 AU max.
- UV Absorbance @ 210 nm:0.04 AU max.
- UV Absorbance @ 225 nm:0.01 AU max.
- UV Absorbance @ 250 nm:0.005 AU max.
- UV Absorbance @ 350 nm:0.005 AU max.
- UV Cutoff:190 nm max.
- Burdick & Jackson™, Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for spectrophotometry:
Specification Test Results
Related Information
Customers Who Bought This Also Bought