Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: VWR International
Certificates
About this item
:67-56-1
:Storage temperature: Ambient
Boiling Pt:64.7 °C (main component)
Melting Pt:-97.68 °C (main component)
Density:0.792 g/cm³ (20 °C. The physical data is that of the main component)
Flash Pt:11 °C
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 64.7 °C (main component)
:Melting Pt:
:Melting Pt: -97.68 °C (main component)
:Flash Pt:
:Flash Pt: 11 °C
:Methanol with 0,1% (v/v) formic acid
:-97.68 °C
:64.7 °C
UN:1230
:3
:II
:11 °C
:GHS02
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):Min. 99.5 %
- Calcium (Ca):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Sodium (Na):Max. 2 ppm
- Transmittance at 254 nm:Min. 90 %
- Transmittance at 230 nm:Min. 10 %
- HPLC Gradient at 254 nm:Max. 10 mAU
- LC-MS application:complying
- Riedel-de Haën™, Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend:
Specification Test Results
Related Information
Customers Who Bought This Also Bought