Guard Column Holders for Metacarb Columns, Agilent Technologies
Catalog #: CAA5204
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Guard Column Holders for Metacarb Columns, Agilent Technologies
Catalog #: CAA5204
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Metacarb cartridge holder, 4.6×250 mm
- Cat. no.:CAA5204
Specifications
About this item
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $
Quality Economy Brand Luer Lock Syringes with Attached Needle, Air-Tite
Supplier: Air-Tite
35,02 $ to 279,99 $
Listeria Palcam Selective Enrichment Broth Base Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
663,68 $ to 3 804,95 $