Nebulizers and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent nebulizers and supplies are equivalent to PerkinElmer AA nebulizer components.
- Guaranteed to be compatible with PerkinElmer AA instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Qualified on a genuine PerkinElmer system
- Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
- Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
- Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Nebulizers are a critical part of the Flame AA sample introduction system. Agilent nebulizers for PerkinElmer AA instruments are designed to support a variety of samples and provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
Customers Who Bought This Also Bought