Hydride Generation Supplies for ICP-MS, Agilent Technologies
Catalog #: CAAGG313865128
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Peristaltic pump tubing for ISIS, Ismaprene, 2-stop, orange/purple tabs, 2.54 mm Int. Ø, 1 pack of 6 pcs
- Cat. no.:CAAGG313865128
Specifications
About this item
ISIS Hydride Generation Accessories for analysis of As, Sb, Se, Sn and Hg with ICP-MS systems.
- ISIS hydride generation accessories are specially designed for Agilent instruments to ensure high quality results
- Flexible selection of accessories for every instrument configuration
- Convenient kits and accessories are simple to order and make user maintenance easy to schedule and execute
- Hydride generation offers the best sensitivity for As, Sb, Se, Sn and Hg with ICP-MS, which are all hydride forming elements
Agilent manufactures a range of high quality ICP-MS replacements and consumables including ISIS hydride generation accessories. The range of accessories include PFA, PTFE and pump tubes of various lengths, union and cross joints, a connector assembly with a range of connector options and several additional fixtures and fittings. The ISIS hydride generation accessory is designed to demanding specifications and delivers the market-leading ICP-MS performance that you have come to expect. Maintain system performance and ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.
Customers Who Bought This Also Bought