Graphite Furnace AAS Operating Supplies Kits, Agilent Technologies
Catalog #: CAAG190068000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Zeeman GTA 120 graphite furnace AA operating supplies kit
Specifications
About this item
Typical supplies for Agilent GTA 120 Deuterium and Zeeman graphite furnace AA Spectrometer systems.
- Maintain accurate analysis
- Save as much as 15% over the cost of purchasing supplies individually
- Keep your instruments running for up to 12 months (depending on use) without worrying about reordering supplies
Agilent's spares kit provides the typical operating supplies required for moderate use of the GTA 120 deuterium and Zeeman graphite furnace Atomic Absorption systems.
Customers Who Bought This Also Bought