Flame Atomization System Kits, Agilent Technologies
Catalog #: CAAG190034100
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Mark 7 atomization system operating kit for flame AA
Specifications
About this item
Typical supplies required for up to 12 months use of Agilent flame AA Spectrometer Systems.
- Maintain accurate analysis
- Save as much as 15% over the cost of purchasing supplies individually
- Keep your instruments running for up to 12 months (depending on use) without worrying about reordering supplies
Agilent's spares kit provides the typical operating supplies required for moderate use of Agilent flame Atomic Absorption systems.
Customers Who Bought This Also Bought