Chiller Supplies for ICP-OES, Agilent Technologies
Catalog #: CAAGG183367024
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Strainer for cooling water for Agilent ICP-MS
- Cat. no.:CAAGG183367024
Specifications
About this item
Poly-clear fluid, strainer, and water filters for the ICP-OES G3292A chiller.
- Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
- Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
Ensure that the ICP-OES G3292A chiller is optimally maintained by using the recommended chiller fluid and supplies. Poly-clear plus fluid is comprised of distilled water with algicide and rust inhibitor allowing for optimal chiller performance while the strainer and fluid filter protect particulates from entering the instrument. The chiller coolant levels should be inspected monthly to ensure an appropriate amount of coolant is in the system. Annual maintenance is recommended where the chiller solution should be completely drained and replaced and the strainer and water filter should be cleaned.
Customers Who Bought This Also Bought