Application Kits for MP-AES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
MP-AES application kits for organic solvents, high sensitivity/aqueous samples, and HF acid digests.
- Application specific sample introduction kits allow you to adapt the instrument to handle challenging samples with different applications
- Kits are convenient as they ensure you have all the compatible supplies for these challenging applications
You have a choice of convenient kits for handling challenging samples with the Agilent MP-AES instrument. Kits are available for high sensitivity with aqueous samples, organic solvents and an inert kit recommended for use with hydrofluoric acid digests. Each kit includes the recommended nebulizer or torch, spray chamber, and peristaltic pump tubing for that application.
Customers Who Bought This Also Bought