ACT 80 Supplies, Agilent Technologies
Catalog #: CAAG9910054400
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Quartz cells ACT-80
- Cat. no.:CAAG9910054400
Specifications
About this item
Typical supplies forAgilent ACT 80 Atom Concentrator Tube used with Agilent flame AA Spectrometers.
- Simple, cost effective and offers benefits in low level analyses
- Increases sensitivity by 2 to 3 times for elements determined using the air/acetylene flame
- Improves detection limits by up to 3 fold
- Measurement is as fast as normal flame determinations, and can be used with manual or automated sampling
- Suitable for use with the air-acetylene burner and compatible with any Agilent flame AA instrument
The ACT-80 Atom Concentrator Tube is a quartz tube that is positioned in the air-acetylene flame to enhance the sensitivity of flame AA by 2 or 3 times when compared to that of normal flame AA. An improvement in detection limit is also achieved.
Customers Who Bought This Also Bought