TapeStation Systems Parts and Accessories, Agilent Technologies
Catalog #: CA76508-048
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:96-well sample plates
- Cat. no.:CA76508-048
Specifications
About this item
Accessories and spare parts for the 4150 and 4200 TapeStation systems like plates and foil seals, loading tips, TapeStation Test Tape, Needle Cartridge.
- 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
- Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the TapeStation systems
- TapeStation Test Tape is available for troubleshooting and running System Diagnostics tests
- The Tapes are reusable and expire after two years
- Loading tips are pre-packed and ready to use
Our portfolio of TapeStation systems spare parts and accessories features everything needed to keep your lab productive. Maximize the efficiency of your TapeStation systems today with our genuine Agilent accessories.
The portfolio features additional needle cartridges and TapeStation Test Tape. In addition, if running short on spare parts, extra loading tips are also available, along with 96-well sample plates and foil seals. For enhanced flexibility we also offer 8-well PCR optical tube strips and caps.
Customers Who Bought This Also Bought