Torch Supplies for MP-AES, Agilent Technologies
Catalog #: CAAGG800368003
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Torch cleaning stand, for cleaning Easy-fit MP-AES torches, for 4100/4200 MP-AES
Specifications
About this item
Torch clamps, torch cleaning stand, and inert PTFE ball joint adapter for the MP-AES.
- Torch cleaning stand for the MP-AES torch: Provides a stable platform for suspending the outer tube, intermediate tube, and injector in the acid cleaning solution
- Encloses the container for the acid cleaning solution, reducing the chance of accidental spillage
- Prevents damage to the fragile end of the quartz outer tube of the torch from contact with the base of the container used for the acid cleaning solution
- Reduces premature degradation of the elastomer materials on the torch body as the seal between the torch body and the torch plate limits exposure to acid fumes from the cleaning solution
Agilent MP-AES torch supplies include torch clamps, torch cleaning stand, and PTFE ball joint adaptor for inert MP-AES torch used with the MP-AES instruments.
Customers Who Bought This Also Bought