Tools and Supplies for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Tools for positioning the L'vov platform in the HGA graphite tube, and other supplies PE systems.
- Guaranteed to be compatible with PerkinElmer Graphite Furnace AAS systems
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Qualified on a genuine PerkinElmer system
- Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
- Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
- In stock and available worldwide for 24- to 48-hour delivery (in most regions)
- Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
- Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
These tools and supplies for PerkinElmer graphite furnace AAS systems can assist you with positioning the L'vov platform precisely in the center of the HGA graphite tube. These tools and supplies, include other supplies used with the PerkinElmer Graphite Furnace AA systems.
Customers Who Bought This Also Bought