SPS 4 Well Plate Sampling Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A range of well-plates and supplies to support autosampling from microtiter plates with the SPS 4.
- Well-plate tray adaptor kit, for Agilent SPS 4 Autosampler, locates 96-position well plates on sample tray, requires, but does not, include, the sample probe which is availble seperately
- Eight 96-well microtiter plate capacity, supports up to 768 samples for ICP-MS only
- Adaptor kit to enable sampling from deep well 96 position microtiter well plates, supported when using MassHunter software with Agilent ICP-MS and SPS 4 autosampler
A range of well-plates and supplies are available to support autosampling from microplates with the Agilent SPS 4 autosampler when used with 7700, 7800, 7900, 8800, and 8900 ICP-MS instruments.
The Agilent SPS 4 is used with current ICP-MS instruments, designed to automate the presentation of liquid samples, drawn from tubes held in racks loaded on the rack location mat, to the instrument for analysis. The SPS 4 can accommodate a variety of fixtures, including standards and sample racks, probes and other accessories, including the microtiter well plates.
Customers Who Bought This Also Bought