Other Operating Supplies Kits, Agilent Technologies
Catalog #: CAAG190025200
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:VGA 77 operating supplies kit. Incl. tubing + connectors, peristaltic pump tubes, spare pump beds, gas-liquid separator, mercury flow-through cell, atomization cells + spare hydride module
Specifications
About this item
Typical supplies for up to 12 months use of VGA 77 and SIPS 10/20 used with flame AAS instruments.
- Maintain accurate analysis
- Save as much as 15% over the cost of purchasing supplies individually
- Keep your instruments running for up to 12 months (depending on use) without worrying about reordering supplies
Agilent's spares kit provides the typical operating supplies required for moderate use of the VGA 77 and SIPS 10/20 accessories used with flame Atomic Absorption systems.
Customers Who Bought This Also Bought