Mercury Analyzer Supplies, Agilent Technologies
Catalog #: CAAG9910087000
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Drying tubes, cetac 470419, sp5557
- Cat. no.:CAAG9910087000
Specifications
About this item
Typical supplies available from Agilent to support use of the QuickTrace Mercury Analyzers.
- QuickTrace Mercury Analyzers are easy to maintain with readily accessible consumable items and common service parts
- The commonality of consumable items parts ensures that parts will be available for years to come
- Common supplies available to support the QuickTrace mercury analyzers include: pPatented, non-foaming gas-liquid separator which ensures unprecedented short and long-term precision with fast washout time
- Nafion drying cartridge to remove excess water vapor
The QuickTrace dedicated mercury analyzer is the most stable and sensitive absorbance based system available. Agilent offers a range of supplies to support use of the QuickTrace Mercury Analyzers.
Customers Who Bought This Also Bought