About this item
Ideal for laboratory and industrial applications, our standard and premium Platform Scale features an easy to navigate color touchscreen display and a slim design allowing for stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results.
- The Standard platform scale features multiple weighing units; USB host, USB device or RS232 connectivity; battery or AC power.
- The Premium platform scale includes RS232 interface –allows data connection to PC or printer; AC adapter included; GLP/GMP data – capabilities including date, time, balance serial number, user ID/project ID, and results.
Features also include superior overload protection and expanded connectivity options. Available in standard or premium.