Specifications
- Description:Scalpel blade remover
- Cat. no.:76548-896
Specifications
About this item
The Aunex ABR-100U scalpel blade remover provides a simply safer way of removing scalpel blades. Remove and dispose of used blades with the single-handed blade removal ABR-100U technique. No fingers, forceps, or re-sheathing. No Injuries.
- Simply push and pull to remove scalpel blades without forceps or fingers
- First blade remover to remove larger pathology and autopsy blades
- Removes #40, #60, and #70 blades, in addition to standard blades size 6 to 26
- Remove and dispose of up to 200 blades
The Aunex blade remover deploys the innovative patent pending Uniflex technology that simply lifts and traps the heel of a blade, then safely disengages and removes the blade. No moving parts. No jamming gears or mechanism.