Draining Tray, Polypropylene
Catalog #: 470340-140
Supplier: United Scientific Supplies
Specifications
- Description:Draining tray, polypropylene
- Cat. no.:470340-140
Specifications
About this item
Wide polypropylene draining tray.
- Autoclavable
- Polypropylene
- Wide net-like design
- Tray dimensions: 16×16×4"
This basket, with wide, net like structure on all sizes is useful for draining different laboratory apparatus after washing them. Made of polypropylene, this tray can be repeatedly autoclaved.
Customers Who Bought This Also Bought