Cryo Rack, Polycarbonate
Catalog #: 470339-404
Supplier: United Scientific Supplies
Specifications
- Description:Cryo rack, polycarbonate
- Cat. no.:470339-404
Specifications
About this item
Durable and autoclavable cryo rack.
- Anti-skid rubber feet
- Alpha Numeric grid
- Autoclavable
This durable and autoclavable Cryo rack can hold 50 vials. Unique design at the bottom of each well lets vials be unscrewed without removing them from the rack. The rack features an alpha numeric index, and anti-skid rubber feet.
Customers Who Bought This Also Bought