Specifications
- Capacity:26 L (6.9 gal.)
- Description:Orbital/Linear Shaking Bath, OLS Aqua Pro, 26 L
- Model:OLS 26 L
- Electrical:110/120V, 50/60Hz
- Temperature range:Ambient +5 to 99 °C / 0 to 99 °C with accessory cooling
- Temperature uniformity:±0.1°C (±0.18°F)
- Orbit:9 mm
- Temperature stability:±0.1°C (±0.18°F)
- Ext. W×D×H:33.5×56.5×32.5 cm (13×22×13")
- Heating capacity:1050 W
- Int. W×D×H:23.5×38 cm (9×15"), working area
- Linear speed (strokes/min):20 - 200 strokes/min.
- Linear stroke length:18, 26, 36 mm
- Timer:1 to 999 minutes
- Weight:13.1 kg
- Cat. no.:10158-864
- Supplier No.:OLS26US
Specifications
About this item
Combined orbital and linear shaking mechanism of the OLS26 allows optimisation of aeration and shear forces mixing, for reproducible results.
- Precision digital temperature control
- Temperature range 0 ºC to 99 ºC operation
- Two LED displays (individual displays and controls for temperature and shaking speed)
- Easy changeover from linear to orbital shaking
- Adjustable shaking speed and stroke length
- Universal tray and polycarbonate lid included as standard
- Drain tap for convenient emptying
The OLS26 offers orbital and linear motion in one bath. Simply rotate the tray carrier by 180° to change between the two shaking modes. No need to buy separate trays to achieve orbital or linear motion. OLS26 features a dual display showing temperature and shaking speed and has adjustable shaking speed/stroke length.
Applications:
General use – defrosting, cooling/warming liquids, temperature control of samples
Life-science – microbiological assays, tissue studies, cell cultivation fermentation, bacterial culture, biochemical assays, enzyme assay
Industrial – materials testing, corrosion testing
Biopharm – solubility testing of medical coatings, dissolution, cooling crystallisation
Food & beverage – extractions, food digestion